missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alkaline Phosphatase, Intestinal Antibody (AP-7/6/7), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50102AF647
This item is not returnable.
View return policy
Description
Alkaline Phosphatase, Intestinal Monoclonal antibody specifically detects Alkaline Phosphatase, Intestinal in Bovine samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| Alkaline Phosphatase, Intestinal | |
| Monoclonal | |
| Alexa Fluor 647 | |
| alkaline phosphatase, intestinal, alkaline phosphomonoesterase, EC 3.1.3.1, glycerophosphatase, IAP, Intestinal alkaline phosphatase, intestinal-type alkaline phosphatase, Kasahara isozyme | |
| Calf intestinal alkaline phosphatase | |
| 0.1 mL | |
| Cancer, Ovarian Carcinoma Cell Markers, Stem Cell Markers | |
| 248 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Immunohistochemistry, Immunofluorescence | |
| AP-7/6/7 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Bovine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction