missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alkaline Phosphatase, Liver/Bone/Kidney Antibody (BGN/03/662), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NB100-66385CL1
This item is not returnable.
View return policy
Description
Alkaline Phosphatase, Liver/Bone/Kidney Monoclonal antibody specifically detects Alkaline Phosphatase, Liver/Bone/Kidney in Bacteria,Human samples. It is validated for ELISA,Western Blot
Specifications
| Alkaline Phosphatase, Liver/Bone/Kidney | |
| Monoclonal | |
| CoraFluor 1 | |
| ALPL, BALP | |
| Alkaline phosphatase from human bone | |
| 0.1 mL | |
| Primary | |
| Bacteria, Human | |
| Purified |
| ELISA, Western Blot | |
| BGN/03/662 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 249 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction