missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alkaline Phosphatase / PLAP Mouse, Clone: GM022, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 250-MSM4-P1
This item is not returnable.
View return policy
Specifications
| Alkaline Phosphatase / PLAP | |
| Cytoplasmic, cell surface | |
| GM022 | |
| Unconjugated | |
| NP_001623 | |
| Mouse | |
| 70kDa | |
| Protein A/G Purified | |
| RUO | |
| 250 | |
| IgG2b κ |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% azide | |
| ALPP | |
| Recombinant human PLAP protein | |
| HepG2 cells. Placenta or seminoma | |
| 100 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction