missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alkaline Phosphatase, Tissue Non-Specific Antibody (ALPL/597), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54376B
This item is not returnable.
View return policy
Description
Alkaline Phosphatase, Tissue Non-Specific Monoclonal antibody specifically detects Alkaline Phosphatase, Tissue Non-Specific in Human, Bovine samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Alkaline Phosphatase, Tissue Non-Specific | |
| Monoclonal | |
| Biotin | |
| Alkaline phosphatase liver/bone/kidney isozyme, alkaline phosphatase, liver/bone/kidney, alkaline phosphatase, tissue-nonspecific isozyme, alkaline phosphomonoesterase, APTNAP, AP-TNAP, EC 3.1.3.1, FLJ40094, FLJ93059, glycerophosphatase, HOPS, liver/bone/kidney-type alkaline phosphatase, MGC161443, tissue-nonspecific ALP, TNAP, TNSALPMGC167935 | |
| Recombinant human Alkaline Phosphatase, Tissue Non-Specific protein (Uniprot: P05186) | |
| 100 μg | |
| Stem Cell Markers | |
| 249 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| ALPL/597 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Bovine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction