missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alkaline Phosphatase, Tissue Non-Specific Antibody (SPM372), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47995V
This item is not returnable.
View return policy
Description
Alkaline Phosphatase, Tissue Non-Specific Monoclonal antibody specifically detects Alkaline Phosphatase, Tissue Non-Specific in Human, Bovine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Frozen), Immunofluorescence, CyTOF
Specifications
| Alkaline Phosphatase, Tissue Non-Specific | |
| Monoclonal | |
| DyLight 405 | |
| Alkaline phosphatase liver/bone/kidney isozyme, alkaline phosphatase, liver/bone/kidney, alkaline phosphatase, tissue-nonspecific isozyme, alkaline phosphomonoesterase, APTNAP, AP-TNAP, EC 3.1.3.1, FLJ40094, FLJ93059, glycerophosphatase, HOPS, liver/bone/kidney-type alkaline phosphatase, MGC161443, tissue-nonspecific ALP, TNAP, TNSALPMGC167935 | |
| Bovine alkaline phosphatase (Uniprot: P05186) | |
| 0.1 mL | |
| Cancer, Lipid and Metabolism, Protein Phosphatase, Stem Cell Markers | |
| 249 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunofluorescence | |
| SPM372 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Bovine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction