missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALKBH4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92817-0.1ml
This item is not returnable.
View return policy
Description
ALKBH4 Polyclonal antibody specifically detects ALKBH4 in Human samples. It is validated for Western Blot
Specifications
| ALKBH4 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| ABH4, ALKBH4 alkB, alkylation repair homolog 4 (E. coli) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-170 of human ALKBH4 (NP_060091.1). MAAAAAETPEVLRECGCKGIRTCLICERQRGSDPPWELPPAKTYRFIYCSDTGWAVGTEESDFEGWAFPFPGVMLIEDFVTREEEAELVRLMDRDPWKLSQSGRRKQDYGPKVNFRKQKLKTEGFCGLPSFSREVVRRMGLYPGLEGFRPVEQCNLDYCPERGSAIDPHL | |
| 0.1 mL | |
| Direct Reversal of DNA Damage | |
| 54784 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction