missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ALKBH4 Antibody (SR2366), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27685
This item is not returnable.
View return policy
Description
ALKBH4 Monoclonal antibody specifically detects ALKBH4 in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| ALKBH4 | |
| Monoclonal | |
| Unconjugated | |
| 50 mM Tris-Glycine (pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot | |
| SR2366 | |
| Western Blot 1:500-1:1000, Immunoprecipitation 1:20 | |
| ABH4, ALKBH4 alkB, alkylation repair homolog 4 (E. coli) | |
| Recombinant protein of human ALKBH4 (Uniprot #: Q9NXW9) | |
| 100 μL | |
| Direct Reversal of DNA Damage | |
| 54784 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction