missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ALKBH5 Polyclonal Antibody

Product Code. 15760351 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15760351 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15760351 Supplier Invitrogen™ Supplier No. PA552401

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: EKLKSMTSRD NYKAGSREAA AAAAAAVAAA AAAAAAAEPY PVSGAKRKYQ EDSDPERSDY EEQQLQKEEE ARKVKSGIRQ MRLFSQDECA KIEARIDEVV SRAEKGLYNE HTVD Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 96%.

ALKBH5 (RNA demehtylase ALKBH5) is a dioxygenase that demethylates RNA by oxidative demethylation. Specifically, it demethylates N(6)-methyladenosine (m6A) RNA, the most prevalent internal modification of messenger RNA (mRNA) in higher eukaryotes. Demethylation of m6A mRNA affects mRNA processing and export. ALKBH5 can also demethylate N(6)-methyladenosine in single-stranded DNA (in vitro). It requires molecular oxygen, alpha-ketoglutarate and iron.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALKBH5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ALKBH5
Gene Accession No. Q6P6C2
Gene Alias ABH5; AlkB family member 5, RNA demethylase; alkB homolog 5, RNA demethylase; alkB, alkylation repair homolog 5; alkB, alkylation repair homolog 5 (E. coli); Alkbh5; Alkylated DNA repair protein alkB homolog 5; alpha-ketoglutarate-dependent dioxygenase alkB homolog 5; AW050020; AW215868; E130207K11; OFOXD; OFOXD1; oxoglutarate and iron-dependent oxygenase domain containing; probable alpha-ketoglutarate-dependent dioxygenase ABH5; RGD1309496; RNA demethylase ALKBH5
Gene Symbols ALKBH5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ALKBH5. Recombinant protein control fragment (Product #RP-89694).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54890
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.