missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ALKBH7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110372
This item is not returnable.
View return policy
Description
Immunogen sequence: MAGTGLLALR TLPGPSWVRG SGPSVLSRLQ DAAVVRPGFL STAEEETLSR ELEPELRRRR YEYDHWDAAI HGFRETEKSR WSEASRAILQ RVQAAAFGPG QTLLSSVHVL DLEARGYIKP HVDSIKFCGA TIAGLSLLSP SVMRLVHTQE PGEWLELLLE PGSLYILRGS ARYDFSHEIL RDEESFFGER RIPRGRRISV ICRSLPEGMG PGESGQPPPA C.
ALKBH7 (alkB, alkylation repair homolog 7), also known as SPATA11, is a 221 amino acid protein belonging to the alkB family. ALKBH7 is one of many homologs of the Escherichia coli protein AlkB. AlkB functions to protect DNA and RNA against damage from environmental methylating compounds by directly reversing 1-methyladenine (1-meA) and 3-methylcytosine (3-meC) cytotoxic alkylation lesions in DNA and RNA. The enzyme acts by oxidative demethylation, utilizing ferrous iron and alpha-ketoglutarate as cofactors, 2-oxoglutarate as a co-substrate and molecular oxygen as the oxidizing agent. ALKBH7 is encoded by a gene located on human chromosome 19, which consists of over 63 million bases, houses approximately 1,400 genes and is recognizedfor having the greatest gene density of the human chromosomes.
Specifications
| ALKBH7 | |
| Polyclonal | |
| Unconjugated | |
| ALKBH7 | |
| 2310045B01Rik; 2510008E23Rik; Abh7; alkB homolog 7; alkB, alkylation repair homolog 7; alkB, alkylation repair homolog 7 (E. coli); Alkbh7; Alkylated DNA repair protein alkB homolog 7; alpha-ketoglutarate-dependent dioxygenase alkB homolog 7, mitochondrial; probable alpha-ketoglutarate-dependent dioxygenase ABH7; SPATA11; spermatogenesis associated 11; spermatogenesis cell proliferation related protein; spermatogenesis cell proliferation-related protein; Spermatogenesis-associated protein 11; UNQ6002; UNQ6002/PRO34564 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 66400, 679944, 84266 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.96 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9BT30, Q9D6Z0 | |
| ALKBH7 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-221 of human ALKBH7 (NP_115682.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction