Learn More
Alkyl-DHAP synthase/AGPS Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 21011-1-AP-20UL
 View more versions of this product
Product Code. 16890595
  • € 145.00 / 20µL
Description
The Alkyl-DHAP synthase/AGPS antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human Alkyl-DHAP synthase/AGPS. This antibody recognizes human antigen. The Alkyl-DHAP synthase/AGPS antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
Alkyl-DHAP synthase/AGPS
Polyclonal
Unconjugated
20 μL
8540
IgG
Western Blot, ELISA
560 μg/mL