missing translation for 'onlineSavingsMsg'
Learn More

all-trans-Retinyl acetate, 1.5 million IU/g, in sunflower oil, stabilized, Thermo Scientific Chemicals

Catalog Number p-7077242
Click to view available options
Quantity:
500 g
This item is not returnable. View return policy
127-47-9
QGNJRVVDBSJHIZ-QHLGVNSISA-N
retinyl acetate, vitamin a acetate, retinol acetate, retinol, acetate, all-trans-retinyl acetate, crystalets, vitamin a1 acetate, all-trans-retinol acetate, vitamin a alcohol acetate, davitan a 650
638034
CHEBI:32095
[(2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenyl] acetate
CC1=C(C(CCC1)(C)C)C=CC(=CC=CC(=CCOC(=O)C)C)C
This item is not returnable. View return policy
127-47-9
QGNJRVVDBSJHIZ-QHLGVNSISA-N
retinyl acetate, vitamin a acetate, retinol acetate, retinol, acetate, all-trans-retinyl acetate, crystalets, vitamin a1 acetate, all-trans-retinol acetate, vitamin a alcohol acetate, davitan a 650
638034
CHEBI:32095
[(2E,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethylcyclohexen-1-yl)nona-2,4,6,8-tetraenyl] acetate
CC1=C(C(CCC1)(C)C)C=CC(=CC=CC(=CCOC(=O)C)C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Flash Point >100°C
Molecular Formula C22H32O2
Refractive Index 1.5470 to 1.5550
Quantity 500 g
Merck Index 15,10211
Solubility Information Solubility in water: difficult soluble.
Molecular Weight (g/mol) 328.49
Viscosity 20 mPa.s (60°C)
Formula Weight 328.49
Percent Purity 1.50 to 1.65 million IU/g (Vitamin A) Photometr.
Related Substances 0.14 (A370/A326) max
Chemical Name or Material all-trans-Retinyl acetate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • all-trans-Retinyl acetate, 1.5 million IU/g, in sunflower oil, stabilized
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Reproductive toxicity Category 1B
Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
  • H360D-May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'