Learn More
Brady™ All-weather Vinyl Labels
Make labels that withstand up to 10 years of outdoor abuse with this low-shrink vinyl label material (B-595). Print almost anywhere with a durable Brady™ handheld label maker.
Brand:  Brady™ 170839
 View more versions of this product
Product Code. 17960690
  • € 56.10 / Pack of 1
In Stock
Description
  • Ideal for labeling pipes, equipment, shelves and other industrial surfaces
  • Designed to withstand prolonged exposure to UV light, chemicals, greases and oils, salt spray, cleaners and detergents, and grime
  • Made with high-tack, permanent acrylic adhesive for use on rough, highly-textured, dirty and smooth surfaces alike, especially in cold temperatures
  • Weather-resistant labels designed to last up to 10 years in an outdoor environment
Specifications
Specifications
Not Autoclavable
Permanent Acrylic
Green, White
Continuous
Top
Label
Component and equipment labelling;Electrical labelling;Facility identification;Floor marking;Lean and 5S labelling;Pipe and valve marking;Warehouse marking
-40°C
1 Label(s) / Box
25.40 mm (W) x 7.62 m (L)
-40°C to 82°C
0.102 mm
25 ft.
1 in.
25.40 mm
M4C-1000-595-GN-WT
BS 5609:1986 Section 2 and section 3 Compliant with Directive 2011/65/EU and 2015/863
None
Gloss
Vinyl
Thermal Transfer
Non-sterile
BMP41, BMP51, BMP53, M511, M410, M510
M4C-1000-595-GN-WT
1
-40°F to 180°F
0.004 in
25.40 mm (W) x 7.62 m (L)
7.62 m
25.40 mm
0.5 in.