Learn More
Allergin-1 Antibody (767727) [Janelia Fluor™ 549], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB7638I
Product Code. 18508065
  • ONLINE EXCLUSIVE PRICE
    € 617.00
    € 582.75 / 0.10mL
    Save € 34.25
    5% Off
Description
Allergin-1 Monoclonal antibody specifically detects Allergin-1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Allergin-1
Monoclonal
Janelia Fluor 549
50mM Sodium Borate
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
284021
Store at 4°C in the dark.
IgG1
Flow Cytometry, Immunocytochemistry, CyTOF
767727
Flow Cytometry, Immunocytochemistry, CyTOF-ready
Allergin-1, Allergy Inhibitory Receptor 1, C17orf60, Chromosome 17 Open Reading Frame 60, Mast Cell Antigen 32, Mast Cell Immunoglobulin-Like Receptor 1, MCA32, MCA-32, Probable Mast Cell Antigen 32 Homolog
Mouse myeloma cell line NS0-derived recombinant human Allergin-1, Met1-Lys227, Accession # Q7Z6M3
0.1 mL