missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Allergin-1 Antibody (767727) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB7638JF669
This item is not returnable.
View return policy
Description
Allergin-1 Monoclonal antibody specifically detects Allergin-1 in Human samples. It is validated for Flow Cytometry,Immunocytochemistry,CyTOF
Specifications
| Allergin-1 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| Allergin-1, Allergy Inhibitory Receptor 1, C17orf60, Chromosome 17 Open Reading Frame 60, Mast Cell Antigen 32, Mast Cell Immunoglobulin-Like Receptor 1, MCA32, MCA-32, Probable Mast Cell Antigen 32 Homolog | |
| Mouse myeloma cell line NS0-derived recombinant human Allergin-1, Met1-Lys227, Accession # Q7Z6M3 | |
| RUO | |
| 284021 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 767727 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur