missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Allophycocyanin Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB8927R-100UG
This item is not returnable.
View return policy
Description
Allophycocyanin Monoclonal specifically detects Allophycocyanin in Multi-Species samples. It is validated for Flow Cytometry.
Specifications
| Allophycocyanin | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Supplied in a saline solution containing BSA and Sodium Azide. | |
| APC | |
| Allophycocyanin isolated from red algae Accession # P25054 | |
| 100 μg | |
| Primary | |
| Reconstitution default not known | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. |
| Flow Cytometry | |
| 936809 | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P25054 | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Detects Allophycocyanin in flow cytometry. | |
| Multi-species | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction