missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Allophycocyanin Alexa Fluor 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB8927N-100UG
This item is not returnable.
View return policy
Description
Allophycocyanin Monoclonal specifically detects Allophycocyanin in Multi-Species samples. It is validated for Flow Cytometry.
Specifications
| Allophycocyanin | |
| Monoclonal | |
| Alexa Fluor 700 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| APC | |
| Allophycocyanin isolated from red algae Accession # P25054 | |
| RUO | |
| Detects Allophycocyanin in flow cytometry. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
| Flow Cytometry | |
| 936809 | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| P25054 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Multi-species | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction