missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Allophycocyanin Antibody (936809), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB8927CL1
This item is not returnable.
View return policy
Description
Allophycocyanin Monoclonal antibody specifically detects Allophycocyanin in Broad Species Reactivity samples. It is validated for Flow Cytometry,CyTOF
Specifications
| Allophycocyanin | |
| Monoclonal | |
| CoraFluor 1 | |
| APC | |
| Allophycocyanin isolated from red algae, Accession # P25054 | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
| Flow Cytometry, CyTOF | |
| 936809 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Broad Species Reactivity | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction