missing translation for 'onlineSavingsMsg'
Learn More

Allura Red AC (Technical Grade), TRC

Product Code. 30317050
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30317050 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30317050 Supplier Toronto Research Chemicals Supplier No. TRCA54875025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Allura Red AC (E 129)
CAS 25956-17-6
Molecular Formula C18 H14 N2 O8 S2 . 2 Na
Quantity 25 g
Synonym 2-Naphthalenesulfonic acid, 6-hydroxy-5-[(2-methoxy-5-methyl-4-sulfophenyl)azo]-, disodium salt (9CI), 2-Naphthalenesulfonic acid, 6-hydroxy-5-[(6-methoxy-4-sulfo-m-tolyl)azo]-, disodium salt (8CI), Allura Red 40, Allura Red AC, Allura red, C.I. 16035, C.I. Food Red 17, E 129, E 129 (dye), FD and C Red No. 40, FD&C Red No. 40, FDC Red 40, FDC Red 40 dye, Fancy Red, Food Red 17, Food Red No. 17, Food Red No. 40, Japan Food Red No. 40, Japan Red 40, Red 40, Red No. 40
InChI Formula InChI=1S/C18H16N2O8S2.2Na/c1-10-7-14(16(28-2)9-17(10)30(25,26)27)19-20-18-13-5-4-12(29(22,23)24)8-11(13)3-6-15(18)21;;/h3-9,21H,1-2H3,(H,22,23,24)(H,25,26,27);;/q;2*+1/p-2/b20-19+;;
SMILES [Na+].[Na+].COc1cc(c(C)cc1\N=N\c2c(O)ccc3cc(ccc23)S(=O)(=O)[O-])S(=O)(=O)[O-]
IUPAC Name disodium;6-hydroxy-5-[(E)-(2-methoxy-5-methyl-4-sulfonatophenyl)diazenyl]naphthalene-2-sulfonate
Molecular Weight (g/mol) 496.42
Formula Weight 495.9987
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.