missing translation for 'onlineSavingsMsg'
Learn More

Allyl chloride, 98%, stabilized, Thermo Scientific Chemicals

Catalog Number p-3598298
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
107-05-1, 75-56-9
C3H5Cl
76.53
MFCD00000984
OSDWBNJEKMUWAV-UHFFFAOYSA-N
allyl chloride, 3-chloro-1-propene, 1-propene, 3-chloro, 3-chloropropylene, 3-chloropropene, chlorallylene, allylchlorid, 2-propenyl chloride, 1-chloro-2-propene, allylchloride
7850
CHEBI:82379
3-chloroprop-1-ene
C=CCCl
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
107-05-1, 75-56-9
C3H5Cl
76.53
MFCD00000984
OSDWBNJEKMUWAV-UHFFFAOYSA-N
allyl chloride, 3-chloro-1-propene, 1-propene, 3-chloro, 3-chloropropylene, 3-chloropropene, chlorallylene, allylchlorid, 2-propenyl chloride, 1-chloro-2-propene, allylchloride
7850
CHEBI:82379
3-chloroprop-1-ene
C=CCCl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point -136.0°C
Color Colorless to Yellow
Density 0.9390g/mL
Boiling Point 44.0°C to 46.0°C
Flash Point −29°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CHCH2Cl
Refractive Index 1.4140 to 1.4160
Quantity 100 mL
Beilstein 01,198
Fieser 13,198
Merck Index 15,28
Specific Gravity 0.939
Solubility Information Solubility in water: 3.6g/L (20°C). Other solubilities: soluble in methanol,ethanol,2-propanol>50g/100mL,soluble in ch2cl2,acetone >50g/100 mL,soluble in n,n-dimethylformamide >50g/100 mL,miscible with chloroform and ether
Stabilizer 0.05 to 0.09% propylene oxide
Viscosity 0.34 mPa.s (20°C)
Formula Weight 76.53
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Allyl chloride
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Allyl chloride, stabilized
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H351-Suspected of causing cancer.
  • H373-May cause damage to organs.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P403+P235-Store in a well-ventilated place. Keep cool.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'