missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Almitrine Bismesylate, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | Almitrine Bismesylate |
| CAS | 29608-49-9 |
| Molecular Formula | C28H37F2N7O6S2 |
| Quantity | 10 g |
| Synonym | 6-[4-[Bis(4-fluorophenyl)methyl]-1-piperazinyl]-N,N'-di-2-propenyl-1,3,5-triazine-2,4-diamine, Dimethanesulfonate (9CI), 2,4-Bis(allylamino)-6-[4-[bis(p-fluorophenyl)methyl]-1-piperazinyl]-s-triazine, dimethanesulfonate (8CI), 1-(4,6-Bis(allylamino)-2-s-triazinyl)-4-(4,4'- difluorobenzhydryl)piperazine bis(methane sulfonate), 1-(4,6-Diallylamino-2-triazinyl)-4-(di-p-fluorobenzhydryl)piperazine bis(methanesulfonate), 1-[4,6- Bis(allylamino)-s-triazin-2-yl]-4-(di-p-fluorobenzhydryl)piperazine bis(methanesulfonate), Almitrine Bismethanesulfonate, Almitrine Dimesylate, Almitrine Dimethanesulfonate, Almitrine Mesylate, Vectarion |
| InChI Formula | InChI=1S/C26H29F2N7.2CH4O3S/c1-3-13-29-24-31-25(30-14-4-2)33-26(32-24)35-17-15-34(16-18-35)23(19-5-9-21(27)10-6-19)20-7-11-22(28)12-8-20;2*1-5(2,3)4/h3-12,23H,1-2,13-18H2,(H2,29,30,31,32,33);2*1H3,(H,2,3,4) |
| SMILES | F[C](F)F.O=[C]O.O=C([C@H](CC1=CN=CN1)NC(C2NC(CC2)=O)=O)N[C@H](C(N[C@@H](CO)C(N[C@H](CC3=CC=C(O)C=C3)C(N[C@@H](CC(C)C)C(NC(C(C([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])([2H])[2H])([2H])C(NC(CCCNC(N)=N)C(N4CCC[C@@H]4C(NCC)=O)=O)=O)=O)=O)=O)=O)CC5=CNC6=CC=CC=C65 |
| IUPAC Name | 6-[4-[bis(4-fluorophenyl)methyl]piperazin-1-yl]-2-N,4-N-bis(prop-2-enyl)-1,3,5-triazine-2,4-diamine;methanesulfonic acid |
| Molecular Weight (g/mol) | 669.76 |
| Formula Weight | 669.2215 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction