Learn More
Almitrine mesylate, MedChemExpress
Product Code. 16465076
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Product Code. 16465076
Brand: Medchem Express HY107319
MedChemExpress Almitrine mesylate, a peripheral chemoreceptor agonist, inhibits selectively the Ca2+-dependent K+ channel.
Almitrine mesylate, a peripheral chemoreceptor agonist, inhibits selectively the Ca2+-dependent K+ channel.
Specifications
Chemical Name or Material Almitrine mesylate
CAS 29608-49-9
Color White
Purity Grade Notes Research
Molecular Formula C28H37F2N7O6S2
Quantity 100 mg
Synonym Almitrine bismesylate Almitrine bismethanesulfonate Almitrine dimesylate
Solubility Information DMSO : 100 mg/mL (149.31 mM; Need ultrasonic)
SMILES C=CCNC1=NC(N(CC2)CCN2C(C3=CC=C(F)C=C3)C4=CC=C(F)C=C4)=NC(NCC=C)=N1.O=S(O)(C)=O.O=S(O)(C)=O
Molecular Weight (g/mol) 669.76
Formula Weight 669.76
Percent Purity 99.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less
Research purposes only