Learn More
IKA™ Alnico V Magnetic Stirring Bars
Shop All IKA Products
Click to view available options
Length (Metric):
10 mm
15 mm
25 mm
30 mm
50 mm
For Use With (Equipment):
Flat bottom containers
Mixing Reagents
Unit Size:
Each
Pack of 5
Specifications
Specifications
| For Use With (Equipment) | Flat bottom containers |
| Diameter (Metric) | 8 mm |
| Length (Metric) | 50 mm |
| Material | Alnico Magnet |
| Quantity | 5/Pk. |
| Shape | Sphere, Cylinder |