Learn More
Alnustone, TRC
Specifications
Specifications
| Chemical Name or Material | (4E,6E)-1,7-diphenylhepta-4,6-dien-3-one |
| CAS | 33457-62-4 |
| Molecular Formula | C19 H18 O |
| Quantity | 25 mg |
| Synonym | (4E,6E)-1,7-Diphenyl-4,6-heptadien-3-one, (E,E)-1,7-Diphenyl-4,6-heptadien-3-one |
| InChI Formula | InChI=1S/C19H18O/c20-19(16-15-18-11-5-2-6-12-18)14-8-7-13-17-9-3-1-4-10-17/h1-14H,15-16H2/b13-7+,14-8+ |
| SMILES | O=C(CCc1ccccc1)\C=C\C=C\c2ccccc2 |
| IUPAC Name | (4E,6E)-1,7-diphenylhepta-4,6-dien-3-one |
| Molecular Weight (g/mol) | 262.346 |
| Formula Weight | 262.136 |