missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALOX5 Monoclonal Antibody (OTI2B7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA807020S
This item is not returnable.
View return policy
Description
ALOX5 Monoclonal antibody specifically detects ALOX5 in Human samples. It is validated for Western BlotSpecifications
| ALOX5 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P15311 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 7430 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI2B7 | |
| Unconjugated | |
| Ezr | |
| 5 - Lipoxygenase; 5-lipoxygenase; 5LO; 5-LO; 5-LOX; 5LPG; 5LX; AI850497; ALOX5; Arachidonate 5-lipoxygenase; arachidonic 5-lipoxygenase alpha-10 isoform; arachidonic 5-lipoxygenase delta-10-13 isoform; arachidonic 5-lipoxygenase delta-13 isoform; arachidonic 5-lipoxygenase delta-p10 isoform; arachidonic acid 5-lipoxygenase; F730011J02; leukotriene A4 synthase; LOG5; LOW QUALITY PROTEIN: arachidonate 5-lipoxygenase; lox5; LOX-5; LOX5A; polyunsaturated fatty acid 5-lipoxygenase; RP11-67C2.3 | |
| Full length human recombinant protein of human EZR produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction