Learn More
ALOX5 Monoclonal Antibody (OTI2D7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA807021
Encompass_Preferred
 View more versions of this product
Product Code. 17547353
  • € 540.87 / 100µL
Description
The enzyme 5-lipoxygenase (5-LO) plays a key role in regulating the production of leukotrienes (LT ). Overproduction of LT contributes to several diseases, most notably chronic inflammatory diseases, including asthma, fibrosis and atherosclerosis. Recent work has demonstrated that the activity of 5-LO is regulated by PKA phosphorylation of serine-523 in 5-LO. Under normal conditions, this phosphorylation may be important in limiting inflammation. Abnormal signaling through cAMP and PKA, then, could contribute to a variety of diseases, including those characterized by chronic inflammation.
Specifications
Specifications
ALOX5
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P09917
ALOX5
Human recombit protein fragment corresponding to amino acids 18-364 of human ALOX5 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Western Blot
OTI2D7
Unconjugated
ALOX5
5-LO, 5-LOX, 5LPG, LOG5
Mouse
Affinity Chromatography
RUO
240
-20° C, Avoid Freeze/Thaw Cycles