missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ alpha 1-Microglobulin Antibody (AMBP/4533) [DyLight 594], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-26935DL594
This item is not returnable.
View return policy
Description
alpha 1-Microglobulin Monoclonal antibody specifically detects alpha 1-Microglobulin in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| alpha 1-Microglobulin | |
| Monoclonal | |
| DyLight 594 | |
| A1M, alpha-1-microglobulin/bikunin precursor, bikunin, EDC1, HCPcomplex-forming glycoprotein heterogeneous in charge, HI30, IATIL, inter-alpha-trypsin inhibitor light chain, ITI, ITILC, ITILgrowth-inhibiting protein 19, protein AMBP, protein HC, trypstatin, uristatin, uronic-acid-rich protein, UTI | |
| Recombinant fragment (around aa200-353) of human alpha 1-Microglobulin protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Cancer | |
| 259 | |
| Store at 4°C in the dark. | |
| IgG2 κ |
| Immunohistochemistry (Paraffin) | |
| AMBP/4533 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering