missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ alpha 1-Microglobulin Antibody (AMBP/4533), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-26787-100ug
This item is not returnable.
View return policy
Description
alpha 1-Microglobulin Monoclonal antibody specifically detects alpha 1-Microglobulin in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| alpha 1-Microglobulin | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT | |
| A1M, alpha-1-microglobulin/bikunin precursor, bikunin, EDC1, HCPcomplex-forming glycoprotein heterogeneous in charge, HI30, IATIL, inter-alpha-trypsin inhibitor light chain, ITI, ITILC, ITILgrowth-inhibiting protein 19, protein AMBP, protein HC, trypstatin, uristatin, uronic-acid-rich protein, UTI | |
| Recombinant fragment (around aa200-353) of human alpha 1-Microglobulin protein (exact sequence is proprietary) | |
| 100 μg | |
| Cancer | |
| 259 | |
| Store at 4°C. | |
| IgG2 κ |
| Immunohistochemistry (Paraffin) | |
| AMBP/4533 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction