missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-1B Adrenergic R/ADRA1B Antibody (471802), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB4730T
This item is not returnable.
View return policy
Description
alpha-1B Adrenergic R/ADRA1B Monoclonal antibody specifically detects alpha-1B Adrenergic R/ADRA1B in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| alpha-1B Adrenergic R/ADRA1B | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 471802 | |
| Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| ADRA1, adrenergic, alpha-1B-, receptor, alpha-1B adrenergic receptor, Alpha-1B adrenoceptor, Alpha-1B adrenoreceptor, alpha-1B-adrenergic receptor, ALPHA1BAR | |
| NS0 mouse myeloma cell line transfected with human ADRA1B, Met1-Phe520, Accession # P35368 | |
| 0.1 mL | |
| GPCR, Neuroscience | |
| 147 | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction