missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-1D Adrenergic R/ADRA1D Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97580AF532
This item is not returnable.
View return policy
Description
alpha-1D Adrenergic R/ADRA1D Polyclonal specifically detects alpha-1D Adrenergic R/ADRA1D in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| alpha-1D Adrenergic R/ADRA1D | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| ADRA1, ADRA1ADAR, ADRA1R, adrenergic, alpha -1D-, receptor, adrenergic, alpha-1A-, receptor, adrenergic, alpha-1D-, receptor, ALPHA1, Alpha-1A adrenergic receptor, alpha-1D adrenergic receptor, Alpha-1D adrenoceptor, Alpha-1D adrenoreceptor, alpha-1D-adrenergic receptor, Alpha-adrenergic receptor 1a, dJ779E11.2 | |
| Produced in rabbits immunized with E. coli-derived Human alpha-1D Adrenergic R/ADRA1D fragment. | |
| 0.1 mL | |
| Cell Cycle and Replication, GPCR, Neuroscience | |
| 146 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction