missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha 2-Macroglobulin Antibody (134), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-89720CL1
This item is not returnable.
View return policy
Description
alpha 2-Macroglobulin Monoclonal antibody specifically detects alpha 2-Macroglobulin in Human samples. It is validated for ELISA
Specifications
| alpha 2-Macroglobulin | |
| Monoclonal | |
| CoraFluor 1 | |
| alpha-2-M, alpha-2-macroglobulin, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 5, CPAMD5DKFZp779B086, FWP007, S863-7 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human alpha 2-Macroglobulin (Accession#: NP_000005.2; Met1-Ala1474). | |
| 0.1 mL | |
| Neuroscience | |
| 2 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| ELISA | |
| 134 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction