missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha 2-Macroglobulin Antibody (A2M/3623), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08689B
This item is not returnable.
View return policy
Description
alpha 2-Macroglobulin Monoclonal antibody specifically detects alpha 2-Macroglobulin in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein Array
Specifications
| alpha 2-Macroglobulin | |
| Monoclonal | |
| Biotin | |
| alpha-2-M, alpha-2-macroglobulin, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 5, CPAMD5DKFZp779B086, FWP007, S863-7 | |
| Human recombinant alpha 2-Macroglobulin protein fragment (around aa 604-748) (exact sequence is proprietary) (Uniprot: P01023) | |
| 100 μg | |
| Neuroscience | |
| 2 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin), Protein Array | |
| A2M/3623 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction