Learn More
alpha 2-Macroglobulin Antibody (A2M/4848), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-20831AF594
Product Code. 18567127
  • € 762.00 / 0.10mL
Description
alpha 2-Macroglobulin Monoclonal antibody specifically detects alpha 2-Macroglobulin in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
alpha 2-Macroglobulin
Monoclonal
Alexa Fluor 594
alpha-2-M, alpha-2-macroglobulin, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 5, CPAMD5DKFZp779B086, FWP007, S863-7
Human recombinant alpha 2-Macroglobulin protein fragment (around aa 604-748) (exact sequence is proprietary)
0.1 mL
Neuroscience
2
Store at 4°C in the dark.
IgG
Immunohistochemistry (Paraffin)
A2M/4848
50 mM Sodium Borate
Mouse
Protein A or G purified