missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha 2-Macroglobulin-like 1/A2ML1 Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00144568-B01P
This item is not returnable.
View return policy
Description
alpha 2-Macroglobulin-like 1/A2ML1 Polyclonal antibody specifically detects alpha 2-Macroglobulin-like 1/A2ML1 in Human, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| alpha 2-Macroglobulin-like 1/A2ML1 | |
| Polyclonal | |
| Western Blot 1:500, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence | |
| AAH93840.1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 144568 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.4) | |
| alpha-2-macroglobulin-like 1, alpha-2-macroglobulin-like protein 1, C3 and PZP-like, alpha-2-macroglobulin domain containing 9 | |
| A2ML1 (AAH93840.1, 1 a.a. - 158 a.a.) full-length human protein. MYTLEASGQGCVYVQTVLRYNILPPTNMKTFSLSVEIGKARCEQPTSPRSLTLTIHTSYVGSRSSSNMAIVEVKMLSGFSPMEGTNQLLLQQPLVKKVEFGTDTLNIYLDELIKNTQTYTFTISQSVLVTNLKPATIKVYDYYLPDEQATIQYSDPCE | |
| 0.05 mg | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu