missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Actinin 1 Antibody (OTI7A4), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70078F
This item is not returnable.
View return policy
Description
alpha-Actinin 1 Monoclonal specifically detects alpha-Actinin 1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockout Validated.
Specifications
| alpha-Actinin 1 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| ACTN1 | |
| Recombinant protein fragment corresponding to amino acids 650-893 of human alpha-Actinin 1 (NP_001093) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2a |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoassay | |
| OTI7A4 | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockout Validated | |
| actinin 1 smooth muscle, actinin, alpha 1, alpha-actinin 1, Alpha-actinin cytoskeletal isoform, alpha-actinin-1, F-actin cross-linking protein, FLJ40884, FLJ54432, Non-muscle alpha-actinin-1 | |
| Mouse | |
| Affinity Purified | |
| Angiogenesis, Cancer, Cytoskeleton Markers | |
| 87.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction