missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alpha Actinin 2 Antibody (ACTN2/3291), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08584AF647
This item is not returnable.
View return policy
Description
Alpha Actinin 2 Monoclonal antibody specifically detects Alpha Actinin 2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein Array
Specifications
| Alpha Actinin 2 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| actinin, alpha 2, alpha-actinin skeletal muscle, Alpha-actinin skeletal muscle isoform 2, alpha-actinin-2, CMD1AA, F-actin cross-linking protein | |
| A recombinant fragment (aa557-692) of human Alpha Actinin 2 protein (exact sequence is proprietary) (Uniprot: P35609) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin), Protein Array | |
| ACTN2/3291 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 88 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction