Learn More
Alpha Actinin 2 Antibody (ACTN2/3293), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-79839AF350
Product Code. 18222537
  • € 635.00 / 0.10mL
Description
Alpha Actinin 2 Monoclonal specifically detects Alpha Actinin 2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Alpha Actinin 2
Monoclonal
Alexa Fluor 350
50mM Sodium Borate with 0.05% Sodium Azide
ACTN2
A recombinant fragment (aa557-692) of human Alpha Actinin 2 protein (exact sequence is proprietary) (Uniprot: P35609)
0.1 mL
Primary
Store at 4°C in the dark.
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
ACTN2/3293
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
actinin, alpha 2, alpha-actinin skeletal muscle, Alpha-actinin skeletal muscle isoform 2, alpha-actinin-2, CMD1AA, F-actin cross-linking protein