missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alpha Actinin 2 Mouse anti-Human, Clone: ACTN2/3291, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07465-100ug
This item is not returnable.
View return policy
Beschreibung
Alpha Actinin 2 Monoclonal specifically detects Alpha Actinin 2 in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.
Spezifikation
| Alpha Actinin 2 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array | |
| actinin, alpha 2, alpha-actinin skeletal muscle, Alpha-actinin skeletal muscle isoform 2, alpha-actinin-2, CMD1AA, F-actin cross-linking protein | |
| A recombinant fragment (aa557-692) of human Alpha Actinin 2 protein(exact sequence is proprietary) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| ELISA, Peptide Array | |
| ACTN2/3291 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 88 | |
| Store at 4°C. | |
| IgG1 κ |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur