missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alpha Actinin 2 Mouse anti-Human, Clone: ACTN2/3294, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07466-100ug
This item is not returnable.
View return policy
Description
Alpha Actinin 2 Monoclonal specifically detects Alpha Actinin 2 in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin, Protein Array.
Specifications
| Alpha Actinin 2 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 1 to 2 μg/mL, Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array | |
| actinin, alpha 2, alpha-actinin skeletal muscle, Alpha-actinin skeletal muscle isoform 2, alpha-actinin-2, CMD1AA, F-actin cross-linking protein | |
| A recombinant fragment (aa557-692) of human Alpha Actinin 2 protein (exact sequence is proprietary) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry (Paraffin), Peptide Array | |
| ACTN2/3294 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 88 | |
| Store at 4°C. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction