missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alpha Actinin 4, p Tyr4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-23333
This item is not returnable.
View return policy
Description
Alpha Actinin 4 Polyclonal antibody specifically detects Alpha Actinin 4 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Alpha Actinin 4 | |
| Polyclonal | |
| Western Blot 1:1000 | |
| actinin alpha4 isoform, actinin, alpha 4, ACTININ-4, alpha-actinin-4, DKFZp686K23158, F-actin cross-linking protein, focal segmental glomerulosclerosis 1, FSGS, FSGS1, Non-muscle alpha-actinin 4 | |
| Phospho-Alpha Actinin 4 (Tyr-4) synthetic peptide (coupled to carrier protein) corresponding to amino acids in the N-terminus of human alpha actinin 4. This sequence is well conserved in rat and mouse alpha actinin 4, but is not conserved in other alpha actinins. [UniProt# O43707] | |
| 0.1 mL | |
| Phospho Specific | |
| 81 | |
| Store at -20°C. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS, 1 mg/ml BSA and 50% glycerol | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction