missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha Adaptin Antibody (AP6), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB600-1545APC
This item is not returnable.
View return policy
Description
alpha Adaptin Monoclonal specifically detects alpha Adaptin in Human, Mouse, Rat, Bovine, Hamster, Primate samples. It is validated for Flow Cytometry.
Specifications
| alpha Adaptin | |
| Monoclonal | |
| APC | |
| 100 kDa coated vesicle protein A, Adapter-related protein complex 2 alpha-1 subunit, Adaptor protein complex AP-2 subunit alpha-1, adaptor-related protein complex 2, alpha 1 subunit, ADTAAAP2-ALPHA, alpha1-adaptin, Alpha-adaptin A, AP-2 complex subunit alpha-1, CLAPA1adaptin, alpha A, Clathrin assembly protein complex 2 alpha-A large chain, clathrin-associated/assembly/adaptor protein, large, alpha 1, Plasma membrane adaptor HA2/AP2 adaptin alpha A subunit | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 160 | |
| Human, Mouse, Rat, Bovine, Hamster, Primate | |
| Purified |
| Flow Cytometry | |
| AP6 | |
| Flow Cytometry | |
| AP2A1 | |
| Purified adaptors. | |
| 0.1 mL | |
| Primary | |
| Detects assembly polypeptide 2 (AP2) It recognizes the products of both alpha-adaptin genes, alpha A and alpha C as well as an alternatively spliced isoform of alpha A found in neurons. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction