Learn More
alpha-Benzylsuccinic acid, 98+%
Product Code. 11383396
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11383396 250 mg 250mg
11373396 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11383396 Supplier Thermo Scientific Alfa Aesar Supplier No. L12846.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
CAS: 884-33-3 | C11H12O4 | 208.213 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 884-33-3
Molecular Formula C11H12O4
Molecular Weight (g/mol) 208.213
MDL Number MFCD00055798
InChI Key GTOFKXZQQDSVFH-UHFFFAOYSA-N
Synonym 2-benzylsuccinic acid, benzylsuccinic acid, dl-benzylsuccinic acid, alpha-benzylsuccinic acid, d,l-benzylsuccinic acid, phenylmethyl butanedioic acid, beta-carboxybenzenebutanoic acid, butanedioic acid,2-phenylmethyl, .alpha.-benzylsuccinic acid, benzenebutanoic acid, .beta.-carboxy
PubChem CID 3858
ChEBI CHEBI:16054
IUPAC Name 2-benzylbutanedioic acid
SMILES C1=CC=C(C=C1)CC(CC(=O)O)C(=O)O
Specifications
Melting Point 161°C to 163°C
Quantity 250 mg
Beilstein 1966188
Formula Weight 208.22
Percent Purity ≥98%
Chemical Name or Material alpha-Benzylsuccinic acid