missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ alpha Desmuslin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30155
This item is not returnable.
View return policy
Description
alpha Desmuslin Polyclonal antibody specifically detects alpha Desmuslin in Human samples. It is validated for Immunoprecipitation
Specifications
| alpha Desmuslin | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 23336 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Immunoprecipitation | |
| Unconjugated | |
| desmuslin, DMNEC 1.4.99.1, EC 2.6.1.16, KIAA0353synemin beta, SYNDesmuslin, synemin, synemin alpha, synemin, intermediate filament protein | |
| Between 1400 and 1450 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction