missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha Desmuslin Antibody (OTI7H6), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71736AF350
This item is not returnable.
View return policy
Description
alpha Desmuslin Monoclonal specifically detects alpha Desmuslin in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| alpha Desmuslin | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| SYNM | |
| Recombinant protein fragment corresponding to amino acids 329-580 of human alpha Desmuslin (NP_663780) produced in E.coli. | |
| 0.1 mL | |
| 23336.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI7H6 | |
| Western Blot, Immunohistochemistry | |
| desmuslin, DMNEC 1.4.99.1, EC 2.6.1.16, KIAA0353synemin beta, SYNDesmuslin, synemin, synemin alpha, synemin, intermediate filament protein | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction