missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ alpha Dystroglycan Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116504
This item is not returnable.
View return policy
Description
Positive Samples: HepG2, U2OS, SW480, HeLa, mouse skeletal muscle, mouse lung, mouse heart, rat liver, rat lung Immunogen sequence: RKKRKGKLTL EDQATFIKKG VPIIFADELD DSKPPPSSSM PLILQEEKAP LPPPEYPNQS VPETTPLNQD TMGEYTPLRD EDPNAPPYQP PPPFTAPMEG KGSRPKNMTP YRSPPPYVPP.
Dystroglycan is a laminin binding component of the dystrophin-glycoprotein complex which provides a linkage between the subsarcolemmal cytoskeleton and the extracellular matrix. Dystroglycan 1 is a candidate gene for the site of the mutation in autosomal recessive muscular dystrophies. The dramatic reduction of dystroglycan 1 in Duchenne muscular dystrophy leads to a loss of linkage between the sarcolemma and extracellular matrix, rendering muscle fibers more susceptible to necrosis. Dystroglycan also functions as dual receptor for agrin and laminin-2 in the Schwann cell membrane. The muscle and nonmuscle isoforms of dystroglycan differ by carbohydrate moieties but not protein sequence. Alternative splicing results in multiple transcript variants all encoding the same protein.
Specifications
| alpha Dystroglycan | |
| Polyclonal | |
| Unconjugated | |
| DAG1 | |
| 156DAG; A3a; AGRNR; Alpha-DG; Alpha-dystroglycan; Beta-DG; Beta-dystroglycan; D9Wsu13e; DAG; DAG1; Dag-1; DG; Dp427; Dp71; dystroglycan; dystroglycan 1; dystroglycan 1 (dystrophin-associated glycoprotein 1); dystrophin associated glycoprotein 1; Dystrophin-associated glycoprotein 1; MDDGA9; MDDGC7; MDDGC9 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 114489, 13138 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 0.87 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q62165 | |
| DAG1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 776-895 of human DAG1 (NP_0011711081). | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction