Learn More
alpha-Fetoprotein/AFP Antibody (189502), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB1368V
Product Code. 30066383
  • € 652.00 / 0.10mL
Description
alpha-Fetoprotein/AFP Monoclonal antibody specifically detects alpha-Fetoprotein/AFP in Human, Mouse samples. It is validated for Western Blot, Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF, KnockDown
TRUSTED_SUSTAINABILITY
Specifications
Specifications
alpha-Fetoprotein/AFP
Monoclonal
Alexa Fluor 405
50mM Sodium Borate
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human, Mouse
Purified
Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF
189502
Western Blot, Simple Western, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready, Knockout Validated
Alpha-1-fetoprotein, Alpha-fetoglobulin, alpha-fetoprotein, FETA, HP, HPAFP
Human umbilical cord serum-derived alpha-Fetoprotein/AFP
0.1 mL
Cancer, Cancer Stem Cells, Developmental Biology, GPCR, Stem Cell Markers, Tumor Biomarkers
174
Store at 4C in the dark.