missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Fetoprotein/AFP Antibody (189508), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals MAB13692C
This item is not returnable.
View return policy
Description
alpha-Fetoprotein/AFP Monoclonal specifically detects alpha-Fetoprotein/AFP in Human samples. It is validated for Western Blot, Knockout Validated.
Specifications
| alpha-Fetoprotein/AFP | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| AFP | |
| Human umbilical cord serum-derived alpha -Fetoprotein/AFP | |
| 100 μL | |
| Cancer, Cancer Stem Cells, GPCR, Stem Cell Markers, Tumor Biomarkers | |
| 174 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot | |
| 189508 | |
| Western Blot | |
| Alpha-1-fetoprotein, Alpha-fetoglobulin, alpha-fetoprotein, FETA, HP, HPAFP | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction