missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Fetoprotein/AFP Antibody (OTI2A9), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70103H
This item is not returnable.
View return policy
Description
alpha-Fetoprotein/AFP Monoclonal specifically detects alpha-Fetoprotein/AFP in Human, Canine, Primate, Monkey samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| alpha-Fetoprotein/AFP | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| AFP | |
| Full length recombinant protein of human alpha-Fetoprotein/AFP (NP_001125) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Canine, Primate, Monkey | |
| IgG2b |
| Western Blot, Immunohistochemistry | |
| OTI2A9 | |
| Western Blot, Immunohistochemistry | |
| Alpha-1-fetoprotein, Alpha-fetoglobulin, alpha-fetoprotein, FETA, HP, HPAFP | |
| Mouse | |
| Affinity Purified | |
| Cancer, Cancer Stem Cells, Developmental Biology, GPCR, Stem Cell Markers, Tumor Biomarkers | |
| 174.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction