Learn More
alpha-Fetoprotein/AFP Antibody (SPM334), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-37754MFV500
Product Code. 18786592
  • € 781.00 / 0.10mL
Description
alpha-Fetoprotein/AFP Monoclonal antibody specifically detects alpha-Fetoprotein/AFP in Human, Porcine, Canine, Monkey, Bovine (Negative), Mouse (Negative), Rat (Negative) samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Monoclonal
mFluor Violet 500 SE
Pig