missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alpha Fodrin Antibody (3D7), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-92689G
This item is not returnable.
View return policy
Description
Alpha Fodrin Monoclonal specifically detects Alpha Fodrin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Alpha Fodrin | |
| Monoclonal | |
| DyLight 488 | |
| Q13813 | |
| SPTAN1 | |
| Recombinant construct (expressed in and purified from E. coli) containing the 19th and 20th spectrin repeats and C-terminal EF hand domains of human alpha Fodrin (amino acids 2086-2447) [UniProt# Q13813]. | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 3D7 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| Alpha-II spectrin, EIEE5, FLJ17738, FLJ44613, Fodrin alpha chain, NEAS, spectrin alpha chain, brain, spectrin, alpha, non-erythrocytic 1 (alpha-fodrin), Spectrin, non-erythroid alpha chain, SPTA2 | |
| Mouse | |
| 240 kDa | |
| 0.1 mL | |
| Neuroscience | |
| 6709 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction