missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alpha Fodrin Antibody (S08-7C6), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19965-50ul
This item is not returnable.
View return policy
Description
Alpha Fodrin Monoclonal antibody specifically detects Alpha Fodrin in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Alpha Fodrin | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| S08-7C6 | |
| Unconjugated | |
| Alpha-II spectrin, EIEE5, FLJ17738, FLJ44613, Fodrin alpha chain, NEAS, spectrin alpha chain, brain, spectrin, alpha, non-erythrocytic 1 (alpha-fodrin), Spectrin, non-erythroid alpha chain, SPTA2 | |
| A synthetic peptide of human Alpha Fodrin (Uniprot # Q13813) | |
| 50 μg | |
| Neuroscience | |
| 6709 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction